Order Entry
United States
ContactUsLinkComponent
Toluene-D8, Standard, 2000 μg/mL, Restek
Toluene-D8, Standard, 2000 μg/mL, Restek
Catalog # RK30224
Supplier:  Restek
CAS Number:  
undefined
Toluene-D8, Standard, 2000 μg/mL, Restek
Catalog # RK30224
Supplier:  Restek
Supplier Number:  30224
CAS Number:  

Specifications

  • Pack type:
    Ampule
  • CAS No.:
    2037-26-5
  • Size:
    1 ml
  • Formula:
    C₆D₅CD₃
  • Description:
    Toluene-D8
  • Cat. No.:
    RK30224
  • Supplier no.:
    30224

Specifications

About this item

Solvent packaged in purge-and-trap-grade methanol.

  • 2000 μg/mL in P&T methanol, 1 mL/ampoule

Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.