Order Entry
United States
ContactUsLinkComponent
Chlorobenzene-D5, Standard, 2000 μg/ml, Restek
Chlorobenzene-D5, Standard, 2000 μg/ml, Restek
Catalog # RK30223
Supplier:  Restek
CAS Number:  
undefined
Chlorobenzene-D5, Standard, 2000 μg/ml, Restek
Catalog # RK30223
Supplier:  Restek
Supplier Number:  30223
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Ampule
  • CAS No.:
    3114-55-4
  • Size:
    1 ml
  • Formula:
    C₆D₅Cl
  • Description:
    Chlorobenzene-D5
  • Cat. No.:
    RK30223
  • Supplier no.:
    30223

Specifications

About this item

Solvent packaged in purge-and-trap-grade methanol.

  • 2000 μg/mL in P&T methanol, 1 mL/ampoule

Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.