ContactUsLinkComponent
Semi-quantitative test strips QUANTOFIX Nitrate / Nitrite
Semi-quantitative test strips QUANTOFIX Nitrate / Nitrite
Catalog # 60787-738
Supplier:  Macherey-Nagel
Small Business Enterprise
CAS Number:  
undefined
Semi-quantitative test strips QUANTOFIX Nitrate / Nitrite
Catalog # 60787-738
Supplier:  Macherey-Nagel
Supplier Number:  91313
Small Business Enterprise
CAS Number:  
Specifications
  • Pack type:
    100 Sticks
  • Cat. No.:
    60787-738
  • Supplier no.:
    91313
Specifications
About this item
For semi-quantitative determinations
Gradation: 0 to 500mg/L. Color changes from white to red/violet.