IL-1 beta ELISA MAX™ Deluxe ELISA Kit, BioLegend
432604-BL
:
- Conjugate ELISA:Unconjugated
- Primary antibody reactivity:Hamster
- Target protein:IL-1 beta
- Description:IL-1 beta ELISA MAX™ Deluxe
- Size:5 Plates
- Cat. No.:432604-BL
- Supplier no.:432604
IL-1 beta ELISA MAX™ Deluxe ELISA Kit reacts with Hamster