GM-CSF MAX™ Deluxe ELISA Kit, BioLegend
Catalog # 432204-BL
Supplier: Biolegend
CAS Number:
Specifications
- Conjugate ELISA:Unconjugated
- Primary antibody reactivity:Mouse
- Target protein:GM-CSF
- Description:GM-CSF ELISA MAX™ Deluxe
- Size:5 Plates
- Cat. No.:432204-BL
- Supplier no.:432204
Specifications
About this item
GM-CSF ELISA MAX™ Deluxe ELISA Kit reacts with Mouse