Anti-Salmonella CSA-1 Antibody (AP)
Catalog # 95058-392
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Salmonella CSA-1
- Antigen Symbol:CSA-2
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Salmonella
- Western Blot:Yes
- Size:100 µg
- Cat. No.:95058-392
- Supplier no.:05-91-99
Specifications
About this item
Primary Goat Anti-Salmonella CSA-1 Reacts with Salmonella
Type: Primary
Antigen: Salmonella CSA-1
Clonality: Polyclonal
Clone:
Conjugation: Alkaline Phosphatase
Epitope:
Host: Goat
Isotype:
Reactivity: Salmonella