Anti-Salmonella CSA-1 Antibody (FITC)
Catalog # 95058-190
Supplier: KPL
Specifications
- Antibody Type:Primary
- Antigen Name:Salmonella CSA-1
- Antigen Symbol:CSA-1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Salmonella
- Western Blot:Yes
- Size:500 µg
- Cat. No.:95058-190
- Supplier no.:02-91-99
Specifications
About this item
Primary Goat Anti-Salmonella CSA-1 Reacts with Salmonella
Type: Primary
Antigen: Salmonella CSA-1
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein (FITC)
Epitope:
Host: Goat
Isotype:
Reactivity: Salmonella