Anti-Reni.salmoninarum Antibody (HRP)
Catalog # 95058-340
Supplier: KPL
Specifications
- Antibody Type:Primary
- Antigen Name:R. salmoninarum antigen
- Antigen Symbol:R. salmoninarum
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Renibacterium salmoninarum
- Western Blot:Yes
- Size:100 µg
- Cat. No.:95058-340
- Supplier no.:04-96-91
Specifications
About this item
Primary Goat Anti-Reni.salmoninarum Reacts with Renibacterium salmoninarum
Type: Primary
Antigen: Reni.salmoninarum
Clonality: Polyclonal
Clone:
Conjugation: Horseradish Peroxidase
Epitope:
Host: Goat
Isotype:
Reactivity: Renibacterium salmoninarum