Anti-Reni. salmoninarum Antibody
Catalog # 95058-088
Supplier: KPL
Specifications
- Antibody Type:Primary
- Antigen Name:R. salmoninarum antigen
- Antigen Symbol:R. salmoninarum
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Renibacterium salmoninarum
- Western Blot:Yes
- Size:1 mg
- Cat. No.:95058-088
- Supplier no.:01-96-91
Specifications
About this item
Primary Goat Anti-Reni. salmoninarum Reacts with Renibacterium salmoninarum
Type: Primary
Antigen: Reni. salmoninarum
Clonality: Polyclonal
Clone:
Conjugation: (none)
Epitope:
Host: Goat
Isotype:
Reactivity: Renibacterium salmoninarum