Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
95058-124
: KPL
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Rabbit
- Western Blot:Yes
- Size:500 µg
- Epitope:Heavy and Light chain
- Cat. No.:95058-124
- Supplier no.:02-15-16
Secondary Goat Anti-IgG (H+L) Reacts with Rabbit (Lapine)
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Rabbit