Anti-IgG Goat Polyclonal Antibody (Cy5®)
Catalog # 95058-462
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:Cy5®
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Rabbit
- Western Blot:Yes
- Size:1 mg
- Cat. No.:95058-462
- Supplier no.:072-02-15-06
Specifications
About this item
Secondary Goat Anti-IgG (H+L) Reacts with Rabbit (Lapine)
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit