Order Entry
United States
ContactUsLinkComponent
Anti-IgG gamma GoatPolyclonal Antibody (AP (Alkaline Phosphatase))
Anti-IgG gamma GoatPolyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # 95058-764
Supplier:  KPL
Anti-IgG gamma GoatPolyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # 95058-764
Supplier:  KPL
Supplier Number:  5220-0355

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G gamma chain
  • Antigen Symbol:
    IgG gamma
  • Clonality:
    Polyclonal
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    1 mg
  • Epitope:
    γ chain
  • Cat. No.:
    95058-764
  • Supplier no.:
    0751-1802

Specifications

About this item

Secondary Goat Anti-IgG(gamma) Reacts with Mouse

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: gamma chain
Host: Goat
Isotype:
Reactivity: Mouse