Anti-IgG/IgM Goat Polyclonal Antibody
Catalog # 95058-030
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G And M
- Antigen Symbol:IgG/IgM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Mouse
- Western Blot:Yes
- Size:1 mg
- Epitope:Heavy and Light chain
- Cat. No.:95058-030
- Supplier no.:01-18-09
Specifications
About this item
Secondary Goat Anti-IgG+IgM (H+L) Reacts with Mouse
Type: Secondary
Antigen: IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse