Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 95058-148
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Mouse
- Western Blot:Yes
- Size:500 µg
- Epitope:Heavy and Light chain
- Cat. No.:95058-148
- Supplier no.:02-18-18
Specifications
About this item
Secondary Goat Anti-IgG(H+L)Ab Rb+HSAFITC Reacts with Mouse
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse