Anti-IgM µ Goat Polyclonal Antibody
Catalog # 95058-024
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin M mu-chain
- Antigen Symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Mouse
- Western Blot:Yes
- Size:1 mg
- Epitope:µ-chain
- Cat. No.:95058-024
- Supplier no.:01-18-03
Specifications
About this item
Secondary Goat Anti-IgM (mu) Reacts with Mouse
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Goat
Isotype:
Reactivity: Mouse