Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
95058-222
: KPL
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antibody Fragment:F(c)
- Cat. No.:95058-222
- Supplier no.:04-10-20
Secondary Goat Anti-IgG (Fc) Reacts with Human
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human