Anti-IgM µ Goat Polyclonal Antibody
95058-922
: KPL
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin M mu-chain
- Antigen Symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:1 mg
- Epitope:µ chain
- Antibody Fragment:F(ab')2
- Cat. No.:95058-922
- Supplier no.:201-1003
Secondary Goat Anti-IgM (mu) F(ab')2 frag Reacts with Human
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Human