Order Entry
United States
ContactUsLinkComponent
Anti-IgM µ Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgM µ Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 95059-018
Supplier:  KPL
Anti-IgM µ Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 95059-018
Supplier:  KPL
Supplier Number:  5230-0442

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin M mu-chain
  • Antigen Symbol:
    IgM µ
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    1 mg
  • Epitope:
    µ chain
  • Antibody Fragment:
    F(ab')2
  • Cat. No.:
    95059-018
  • Supplier no.:
    202-1003

Specifications

About this item

Secondary Goat Anti-IgM (µ) F(ab')2, Reacts with Human

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Human