Order Entry
United States
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
  95058-494
 :  KPL
Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
  95058-494
 :  KPL
 :  5230-0382

 

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    1 mg
  • Epitope:
    Heavy and Light chain
  • Cat. No.:
    95058-494
  • Supplier no.:
    072-03-10-06

 

 

Secondary Goat Anti-IgG (H+L) Reacts with Human

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Human