Anti-IgG gamma Goat Polyclonal Antibody (Biotin)
Catalog # 95058-842
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G gamma chain
- Antigen Symbol:IgG gamma
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:500 µg
- Epitope:γ chain
- Cat. No.:95058-842
- Supplier no.:16-10-02
Specifications
About this item
Secondary Goat Anti-IgG (gamma) Reacts with Human
Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: gamma chain
Host: Goat
Isotype:
Reactivity: Human