Anti-IgE epsilon Goat Polyclonal Antibody (Biotin)
Catalog # 95058-846
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin E epsilon
- Antigen Symbol:IgE epsilon
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:500 µg
- Epitope:ε-chain
- Cat. No.:95058-846
- Supplier no.:16-10-04
Specifications
About this item
Secondary Goat Anti-IgE (ε) Reacts with Human
Type: Secondary
Antigen: IgE epsilon
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: epsilon-chain
Host: Goat
Isotype:
Reactivity: Human