Order Entry
United States
ContactUsLinkComponent
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Catalog # 95057-994
Supplier:  KPL
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Catalog # 95057-994
Supplier:  KPL
Supplier Number:  5210-0160

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin A, G And M
  • Antigen Symbol:
    IgA/IgG/IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    1 mg
  • Epitope:
    Heavy and Light chain
  • Cat. No.:
    95057-994
  • Supplier no.:
    01-10-07

Specifications

About this item

Secondary Goat Anti-IgA+IgG+IgM (H+L) Reacts with Human

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Human