Anti-IgA alpha Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # 95058-748
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin A alpha
- Antigen Symbol:IgA alpha
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:1 mg
- Epitope:α chain
- Cat. No.:95058-748
- Supplier no.:0751-1001
Specifications
About this item
Secondary Goat Anti-IgA (α) Reacts with Human
Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: alpha chain
Host: Goat
Isotype:
Reactivity: Human