Order Entry
United States
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 95058-796
Supplier:  KPL
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 95058-796
Supplier:  KPL
Supplier Number:  5220-0371

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Hamster
  • Western Blot:
    Yes
  • Size:
    500 µg
  • Cat. No.:
    95058-796
  • Supplier no.:
    14-22-06

Specifications

About this item

Secondary Goat Anti-IgG (H+L) Reacts with Hamster

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Hamster