Anti-H.pylori Antibody
95058-082
: KPL
- Antibody Type:Primary
- Antigen Name:H. pylori antigen
- Antigen Symbol:H. pylori
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:H. pylori
- Western Blot:Yes
- Size:1 mg
- Cat. No.:95058-082
- Supplier no.:01-93-94
Primary Goat Anti-H. pylori Reacts with Helicobacter pylori
Type: Primary
Antigen: H. pylori
Clonality: Polyclonal
Clone:
Conjugation: (none)
Epitope:
Host: Goat
Isotype:
Reactivity: Helicobacter pylori