Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # 95058-826
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Guinea pig
- Western Blot:Yes
- Size:500 µg
- Epitope:Heavy and Light chain
- Cat. No.:95058-826
- Supplier no.:15-17-06
Specifications
About this item
Secondary Goat Anti-IgG(H+L) Reacts with Guinea Pig
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Guinea Pig