Anti-IgG Rabbit Polyclonal Antibody (Cy5®)
95058-458
: KPL
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:Cy5®
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Goat
- Western Blot:Yes
- Size:1 mg
- Epitope:Heavy and Light chain
- Cat. No.:95058-458
- Supplier no.:072-02-13-06
Secondary Rabbit Anti-IgG (H+L) Reacts with Goat
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat