Anti-O157:H7 Goat Polyclonal Antibody
Catalog # 95058-084
Supplier: KPL
Specifications
- Antibody Type:Primary
- Antigen Name:E. coli O157:H7
- Antigen Symbol:O157:H7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:E. coli
- Western Blot:Yes
- Size:1 mg
- Cat. No.:95058-084
- Supplier no.:01-95-90
Specifications
About this item
Primary Goat Anti-E. coli O157:H7 Reacts with E.coli
Type: Primary
Antigen: O157:H7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: E. coli