Anti-IgM µ Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 95058-244
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin M mu-chain
- Antigen Symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Dog
- Western Blot:Yes
- Size:100 µg
- Epitope:µ-chain
- Cat. No.:95058-244
- Supplier no.:04-19-03
Specifications
About this item
Secondary Goat Anti-IgM (mu) Reacts with Dog (Canine)
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: µ-chain
Host: Goat
Isotype:
Reactivity: Dog