Anti-IgG gamma Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # 95058-350
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G gamma chain
- Antigen Symbol:IgG gamma
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Dog
- Western Blot:Yes
- Size:100 µg
- Epitope:γ chain
- Cat. No.:95058-350
- Supplier no.:051-19-02
Specifications
About this item
Secondary Goat Anti-IgG (g) Reacts with Dog (Canine)
Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: gamma chain
Host: Goat
Isotype:
Reactivity: Dog