Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 95058-776
Supplier: KPL
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Bovine
- Western Blot:Yes
- Size:500 µg
- Epitope:Heavy and Light chain
- Cat. No.:95058-776
- Supplier no.:14-12-06
Specifications
About this item
Secondary Goat Anti-IgG (H+L) Reacts with Cow (Bovine, Cattle)
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Bovine