Order Entry
United States
ContactUsLinkComponent
Anti-B. burgdorferi antigen Goat Polyclonal Antibody
Anti-B. burgdorferi antigen Goat Polyclonal Antibody
Catalog # 95058-090
Supplier:  KPL
Anti-B. burgdorferi antigen Goat Polyclonal Antibody
Catalog # 95058-090
Supplier:  KPL
Supplier Number:  5310-0337

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    B. burgdorferi antigen
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    B. burgdorferi
  • Western Blot:
    Yes
  • Size:
    1 mg
  • Cat. No.:
    95058-090
  • Supplier no.:
    01-97-91

Specifications

About this item

Primary Goat Anti-B. burgdorferi Reacts with Borrelia burgdorferi

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Borrelia burgdorferi