ContactUsLinkComponent
Ratcheting Box Wrench Sets, Proto®, ORS Nasco
  300002-696
undefined
Ratcheting Box Wrench Sets, Proto®, ORS Nasco
  300002-696
 :  577-1190A
  • Material:
    Forged alloy steel
  • Type:
    Ratchet
  • Cat. No.:
    300002-696
  • Weight:
    1.72 lbs.
  • Includes:
    (1) 1/4 x 5/16 in, 4-9/32 in. long, (1) 3/8 x 7/16 in, 5-1/2 in. long, (1) 1/2 x 9/16 in, 6-3/4 in. long, (1) 5/8 x 11/16 in, 8-7/64 in. long, (1) 3/4 x 7/8 in, 9-1/4 in. long, tool roll
  • Supplier no.:
    577-1190A
  • Handle type:
    Flat
  • Measuring system:
    IN
  • No. of points:
    12
Applies more torque to fastener.
  • Reduces risk of fastener rounding
  • Improves grip on worn, rounded fasteners
  • Openings are precision broached