Order Entry
United States
Orders LinkContactUsLinkComponent
Phenol ≥99% ACS

Phenol ≥99% ACS

Toxic
Corrosive
Health
Supplier:   VWR
Phenol ≥99% ACS
Supplier:  VWR
Certificates
Phenol ≥99% ACS

1 Options Available Below

Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Boiling Pt:181.8 °C (1013 hPa)
  • Melting Pt:40.8 °C
  • Density:1.06 g/cm³ (20 °C)
  • Flash Pt:81 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:203-632-7
  • UN:1671
  • ADR:6.1,II

Documents

Specifications for Product Family

Specification Test Results

  • Clarity of Solution:
    Pass
  • Freezing Range:
    =40.5°C
  • Moisture (KF):
    = 0.5%
  • Purity:
    = 99%
  • Residue After Evaporation:
    = 0.05%
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 250 g
    Log in for availability
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $94.33
    $94.33
    Log in for availability
    Case of 12 (250g)
    Case of 12 (250g) - $686.93
    $686.93
    Each (250g)
       $94.33

    Case of 12 (250g)
       $686.93

    Supplier Number: 0735-250G
  • 500 g
    Discontinued
    Supplier Number: -
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.