ContactUsLinkComponent
Industrial Absorbents, Sphag Sorb®, ORS Nasco
Industrial Absorbents, Sphag Sorb®, ORS Nasco
Catalog # 300009-867
undefined
Industrial Absorbents, Sphag Sorb®, ORS Nasco
Catalog # 300009-867
Supplier Number:  671-SS-2B
Specifications
  • Dimensions:
    2 cu. ft.
  • Cat. No.:
    300009-867
  • Packaging:
    Bag, loose-filled
  • Absorbency:
    9 - 12 gal.
  • Weight:
    18 lbs.
  • Supplier no.:
    671-SS-2B
Specifications
About this item
Absorbs fuels, organic chemicals, paints, glycol and more