Orders LinkContactUsLinkComponent
Small Protector Cases, Pelican™
Small Protector Cases, Pelican™
Certificates
Small Protector Cases, Pelican™
Small Protector Cases, Pelican™
2 Options Available Below
Product Details & Documents
These small protector polypropylene polycase is water, dust and crush resistant.
  • Open cell core with solid wall design, strong, light weight
  • Automatic pressure equalization valve
  • Pick N Pluck™ with convoluted lid foam
  • Stainless steel hardware
  • O-ring seal
Temperature range: –40.0 °F [Min], 210.0 °F [Max]
Product Family Options
Product Information
  • Color
    Material
    Weight
    Dimensions
    Availability
    Price
  • Black
    Copolymer Polypropylene
    1.23 lbs.
    6.560×3.58×8.120" (Outer); 4.75×3.06×7.25" (Inner)
    Log in for availability
    Log in for availability
    Each
    Each - $58.16
    $58.16
    Each
       $58.16
    Supplier Number: 562-1120-BLACK
  • Black
    Copolymer Polypropylene
    5.5 lbs.
    11.620×6×13.370" (Outer); 8.87×5.18×11.810" (Inner)
    Log in for availability
    Log in for availability
    Each
    Each - $349.03
    $349.03
    Each
       $349.03
    Supplier Number: 562-1400-BLACK