Medium Protector Cases, Pelican™
Supplier: Safety & Industrial Supplies
Jump to:
About this item
These medium protector polypropylene polycase is water, dust and crush resistant.
- Easy open double throw latches
- Open cell core with solid wall design, strong, light weight
- Automatic pressure equalization valve
- Comfortable rubber over-molded handle
- Stainless steel hardware and padlock protectors
- Pick N Pluck™ with convoluted lid foam
- O-ring seal
Temperature range: −40.0 °F [Min], 210.0 °F [Max]
2 Options Available Below
Product Details & Documents
These medium protector polypropylene polycase is water, dust and crush resistant.
- Easy open double throw latches
- Open cell core with solid wall design, strong, light weight
- Automatic pressure equalization valve
- Comfortable rubber over-molded handle
- Stainless steel hardware and padlock protectors
- Pick N Pluck™ with convoluted lid foam
- O-ring seal
Temperature range: −40.0 °F [Min], 210.0 °F [Max]
Product Family Options
Product Information
- ColorMaterialWeightDimensionsAvailabilityPrice
- BlackCopolymer Polypropylene6.39 lbs.13×6.87×16" (Outer); 10.18×6×14.620" (Inner)Log in for availabilityEach$473.43Supplier Number: 562-1450-BLACK
- BlackCopolymer Polypropylene9.35 lbs.15.770×7.41×19.780" (Outer); 12.89×6.72×18.060" (Inner)Log in for availabilityEach$619.81Supplier Number: 562-1520-BLACK