Respirator Cleaning Pads, Allegro®
Supplier: Safety & Industrial Supplies
Certificates
About this item
Quick and easy answer for cleaning rubber respirator face pieces and other personal safety equipment.
- Non-woven cloths saturated with 70% Isopropanol
- Offered in alcohol free
Packaging: Individually foil-packaged.
Frequently Bought Together