Topgard® Protective Caps and Hats, MSA
Catalog # 300072-588
Supplier: Safety & Industrial Supplies
Specifications
- Color:White
- Description:Fas-Trac® Ratchet
- Cat. No.:300072-588
- Weight:0.89 lbs.
- Supplier no.:454-475385
Specifications
About this item
It is a standard, slotted cap with size group of 6 1/2 - 8
- Uni-ridge design protects wearer from falling objects
- Lightweight and durable
- Limited resistance to top impact or penetration is retained through a wide range of temperatures