Order Entry
United States
ContactUsLinkComponent
Insect Shield™ Insect Repellent Towels, Dymon
Insect Shield™ Insect Repellent Towels, Dymon
Catalog # 300000-304
undefined
Insect Shield™ Insect Repellent Towels, Dymon
Catalog # 300000-304
Supplier Number:  253-91401
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Insecticide against mosquitos black flies, no-see-ums, ticks stable
  • Environmentally Preferable:
  • Cat. No.:
    300000-304
  • Volume:
    4 lbs.
  • Supplier no.:
    253-91401

Specifications

About this item

It is used against mosquitos, black flies, no-see-ums, ticks, stable flies, chiggers, biting flies, fleas, sand flies, deer flies, gnats

  • EPA-approved insect repellant formula combined with a large, lint-free towel
  • With 30% DEET, INSECT SHIELD™ Insect Repellant Towels provide a safe, yet highly effective level of protection
  • Once applied, the INSECT SHIELD™ Insect Repellant formula is waterproof and greaseless