ContactUsLinkComponent
EyeSafe™ Faucet-Mounted Eye Washes, Guardian
EyeSafe™ Faucet-Mounted Eye Washes, Guardian
Catalog # 300068-296
undefined
EyeSafe™ Faucet-Mounted Eye Washes, Guardian
Catalog # 300068-296
Supplier Number:  333-G1100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Cat. No.:
    300068-296
  • Weight:
    1 lb.
  • Supplier no.:
    333-G1100
Specifications
About this item
Converts any faucet into an emergency eye wash station without interfering with normal faucet operation.
  • Adjustable aerated outlet heads
  • Float off dust covers to protect outlet heads
  • Forged brass diverter valve
  • Body has ⁵⁵/₆₄"-27 female thread
  • Furnished with removable aerator on bottom
Pull knob to activate eye wash; water pressure holds eye wash in operation, leaving user's hands free.