Order Entry
United States
ContactUsLinkComponent
Bloodborne Pathogens Kits, Pac-Kit®
Bloodborne Pathogens Kits, Pac-Kit®
Catalog # 300009-477
undefined
Bloodborne Pathogens Kits, Pac-Kit®
Catalog # 300009-477
Supplier Number:  579-3060
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Width:
    7½"
  • Description:
    Blood Borne Pathogen Kit, with Weatherproof Plastic Case
  • Height:
    2¾"
  • Length:
    4½"
  • Environmentally Preferable:
  • Kit Contents:
    Absorbent Powder, 2 oz. Pouch, Antiseptic Towelettes, Biohazard Bag, Clear Bag, Control Gown, X-Large, Eye & Face Shield/Mask, Nitrile Exam Gloves, Paper Towels, Scoop & Spatula, Surface Disinfectant Towelettes
  • Cat. No.:
    300009-477
  • Weight:
    1.75 lbs.
  • Supplier no.:
    579-3060

Specifications

About this item

Supplies to cover just about any workplace injury.

  • Neatly ordered interior compartment
  • Weatherproof case
  • Carry-handle or is wall mountable
Delivery: Includes: Antiseptic towelettes, biohazard bag, clear bag, control gown, X-large, eye and face shield/mask, nitrile exam gloves, paper towels, scoop and spatula, surface disinfectant towelettes.