Altair® Single-Gas Detectors, CO / H2S, MSA
Designed for superior lifespan—patented sensors operate for over two years—and high performance, the ALTAIR Single-Gas Detector is the smart choice for market-leading, cost-effective gas monitors.
- Sensor options for carbon monoxide, hydrogen sulfide, and oxygen
- LED/audible/vibrating alarms
- Superior dust/water protection
- High RFI resistance
- One-button operation provides safety, increased durability and ease-of-use
Frequently Bought Together