Fixed Beam Anchors, DBI-SALA, ORS Nasco
Supplier: Safety & Industrial Supplies
Certificates
About this item
This fixed beam anchor easily clamps to vertical or horizontal I-beams 1¹/₂" thick to provide fixed anchor for lanyards, self-retracting lifelines or pre-engineered horizontal lifelines.
- Also available in models that fit flanges up to 36" wide and 2¹/₂" thick
Specifications
- Catalog No:
- 300058-306
- 300075-620
- Flange Size:
- 2 1/2 in [Min], 12 in [Max]
- 2 1/2 in [Min], 18 in [Max]