Order Entry
United States
ContactUsLinkComponent
 
Howard Leight Laser Trak® Detectable Earplugs, Honeywell Safety
  300000-089
undefined
Howard Leight Laser Trak® Detectable Earplugs, Honeywell Safety
  300000-089
 :  154-LT-30

 

  • Noise reduction rating:
    32 dB
  • Color:
    Orange/Blue
  • Description:
    Corded earplugs
  • Cat. No.:
    300000-089
  • Packaging:
    Poly bag
  • Supplier no.:
    154-LT-30

 

 

These earplugs are made of foam material.

  • Non-ferrous metal grommet and bright colors - easily detected by visual and automated inspection
  • Self-adjusting polyurethane foam expands to fit virtually any wearer
  • Contoured T-shape for easy insertion and wear
  • Maximum attenuation of 52.000 dB at 8000.000 Hz

Weight: 0.01 lbs.

 : Meets ANSI S.3.19-1974 standards.