Mini-Triggertorch™ Microtorch Kits, Master Appliance
About this item
Built-in self-igniting system with safety lock that ensures the switch is not turned on when not in use. Contains butane gas.
- 'Hands-free' slide lock for continuous use and instant off operation
- Durable self-contained, refillable plastic fuel tank
- Gas adjustment wheel provides adjustable, long blue torch flame
- Incorporates 'trigger' switch with 'pulldown' lock to meet Consumer Products Safety Commission (CPSC) Child Safety Requirements
- Self-igniting piezo electric ignition is easy to start and eliminates the need for lighter or matches
Unique shutter lever to easily extinguish ignition flame if using optional soldering or hot air tip attachments.
Ordering information: The promo pack includes MT-11 Microtorch and MT-70 Mini-Triggertorch.
1 Options Available Below
Product Details & Documents
Built-in self-igniting system with safety lock that ensures the switch is not turned on when not in use. Contains butane gas.
- 'Hands-free' slide lock for continuous use and instant off operation
- Durable self-contained, refillable plastic fuel tank
- Gas adjustment wheel provides adjustable, long blue torch flame
- Incorporates 'trigger' switch with 'pulldown' lock to meet Consumer Products Safety Commission (CPSC) Child Safety Requirements
- Self-igniting piezo electric ignition is easy to start and eliminates the need for lighter or matches
Unique shutter lever to easily extinguish ignition flame if using optional soldering or hot air tip attachments.
Ordering information: The promo pack includes MT-11 Microtorch and MT-70 Mini-Triggertorch.
Product Family Options
Product Information
- Operating temperatureWidthLengthHeightWeightAvailabilityPrice
- Supplier Number: 467-MT-70
- 2500 °F [Max]1.300"5½"3.800"¾ lbs.Discontinued-Supplier Number: -