Orders LinkContactUsLinkComponent
Heating Elements, Weller®
900 °F [Max], Thread-on tips, 7400, 7500, 7760, 7770, SL325, SL335, SL345, SL500, 0.07 lbs., 45.00 W
Catalog # 300062-976
 :  185-4037S
Heating Elements, Weller®
Catalog # 300062-976
Heating Elements, Weller®
Heating Elements, Weller®
900 °F [Max], Thread-on tips, 7400, 7500, 7760, 7770, SL325, SL335, SL345, SL500, 0.07 lbs., 45.00 W
Price
$79.87
Log in for availability
Each
Product Details & Documents
Specifications
  • Power:
    45.00 W
  • Weight:
    0.07 lbs.
  • Tip type:
    Thread-on tips
  • Operating temperature:
    900 °F [Max]
  • Use with:
    7400, 7500, 7760, 7770, SL325, SL335, SL345, SL500