ContactUsLinkComponent
Cobalt High Speed Steel Drill Bit Sets, Irwin®, ORS Nasco
Cobalt High Speed Steel Drill Bit Sets, Irwin®, ORS Nasco
Catalog # 300009-512
undefined
Cobalt High Speed Steel Drill Bit Sets, Irwin®, ORS Nasco
Catalog # 300009-512
Supplier Number:  585-3018002
Specifications
  • Cat. No.:
    300009-512
  • Increments:
    ¹⁄₆₄"
  • Type:
    Drill Bit Set
  • Weight:
    4.40 lbs.
  • Includes:
    Metal index case
  • Finish:
    Gold oxide
  • Supplier no.:
    585-3018002
  • Cutting diameter:
    ¹⁄₁₆" (min.), ½" (max.)
Specifications
About this item
Cutting edge material: HSS
  • Cobalt steel extends cutting edge life and maintains cutting life after resharpening
  • 135º split point begins drilling immediately, minimizing bit "walk"
  • Rubber overmolded case protects kit if dropped