Powerlock® Tape Rules 1" Wide Blade, Stanley®
Supplier: Safety & Industrial Supplies
Certificates
About this item
Single side measuring tape in inch in two different lengths.
- Heat treated spring for longer life
- Positive blade lock will not creep while taking measurements
- Entire blade coated with Mylar® Polyester film for added durability
- Belt clip on back of case makes it convenient for carrying around
- Hook at end,
- Spring return motion
ABS case.
Frequently Bought Together