ContactUsLinkComponent
Measure Stix™ Steel Measuring Tapes, L.S. Starrett
Measure Stix™ Steel Measuring Tapes, L.S. Starrett
  300044-082
undefined
Measure Stix™ Steel Measuring Tapes, L.S. Starrett
  300044-082
 :  681-63172
  • Description:
    High carbon steel single side measuring tape, inch and metric
  • Length:
    1.8 m (6 ft.)
  • Environmentally Preferable:
  • Cat. No.:
    300044-082
  • Graduations:
    S12-Inch/mm
  • Width:
    0.75"
  • Supplier no.:
    681-63172
It is high quality precision steel rule is flat with permanent adhesive back.
  • Cut to size with scissors
  • For mounting on workbenches, saw tables, drafting tables, etc.